Anti-TAB2 Rabbit Polyclonal Antibody
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB140201-100EA 6150 DKK
ABCAAB140201-100
Anti-TAB2 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: TAB2 (TGF-beta activated kinase 1/MAP3K7 binding protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human