Anti-TSHZ1 Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB140196-100EA 6630 DKK
ABCAAB140196-100
Anti-TSHZ1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: TSHZ1 (teashirt zinc finger homeobox 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR