Anti-LMX1B Rabbit Polyclonal Antibody
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB139736-100EA 6440 DKK
ABCAAB139736-100
Anti-LMX1B Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: LMX1B (LIM homeobox transcription factor 1, beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat