Anti-ATG14 Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB139727-100EA 6230 DKK
ABCAAB139727-100
Anti-ATG14 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: ATG14 (autophagy related 14)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR