Anti-SMAD3 Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB138659-100EA 4100.24 DKK
ABCAAB138659-100
Anti-SMAD3 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: SMAD3 (SMAD family member 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR