Anti-RRN3 Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB138651-100EA 6480 DKK
ABCAAB138651-100
Anti-RRN3 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: RRN3 (RRN3 RNA polymerase I transcription factor homolog (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR