Anti-CYP7B1 Rabbit Monoclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB138497-100EA 6620 DKK
ABCAAB138497-100
Anti-CYP7B1 Rabbit Monoclonal Antibody
Antistoffer
Type: Primary
Antigen: CYP7B1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR