Anti-E. coli Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB137967-500EA 4286.61 DKK
ABCAAB137967-500
Anti-E. coli Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: E. coli
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: E. coli
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR