Anti-POM121 Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB137938-100EA 6480 DKK
ABCAAB137938-100
Anti-POM121 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: POM121 (POM121 transmembrane nucleoporin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR