Anti-PIKFYVE Rabbit Polyclonal Antibody
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB137907-100EA 6150 DKK
ABCAAB137907-100
Anti-PIKFYVE Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: PIKFYVE (phosphoinositide kinase, FYVE finger containing)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human