Anti-SRPK3 Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB137813-100EA 3802.04 DKK
ABCAAB137813-100
Anti-SRPK3 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: SRPK3 (SRSF protein kinase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR