Anti-HSPA13 Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB137680-100EA 6580 DKK
ABCAAB137680-100
Anti-HSPA13 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: HSPA13 (heat shock protein 70kDa family, member 13)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR