Anti-DUSP2 Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB137640-100EA 6380 DKK
ABCAAB137640-100
Anti-DUSP2 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: DUSP2 (dual specificity phosphatase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR