Anti-LMO1 Rabbit Polyclonal Antibody
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB137599-50EA 5890 DKK
ABCAAB137599-50
Anti-LMO1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: LMO1 (LIM domain only 1 (rhombotin 1))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse