Anti-CBLB Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB137596-50EA 4098.45 DKK
ABCAAB137596-50
Anti-CBLB Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: CBLB (Cbl proto-oncogene B, E3 ubiquitin protein ligase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR