Anti-BCAR1 Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB137507-50EA 4172.96 DKK
ABCAAB137507-50
Anti-BCAR1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: BCAR1 (breast cancer anti-estrogen resistance 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR