Anti-HUNK Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB137492-100EA 6150 DKK
ABCAAB137492-100
Anti-HUNK Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: HUNK (hormonally up-regulated Neu-associated kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR