Anti-P2RY13 Rabbit Polyclonal Antibody
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB137481-100EA 3949.41 DKK
ABCAAB137481-100
Anti-P2RY13 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: P2RY13 (purinergic receptor P2Y, G-protein coupled, 13)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human