Anti-PSMC5 Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB137476-100EA 4098.45 DKK
ABCAAB137476-100
Anti-PSMC5 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: PSMC5 (proteasome (prosome, macropain) 26S subunit, ATPase, 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR