Anti-TRAF6 Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB137452-100EA 6650 DKK
ABCAAB137452-100
Anti-TRAF6 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: TRAF6 (TNF receptor-associated factor 6, E3 ubiquitin protein ligase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR