Anti-PIP5K1A Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB137441-100EA 0 DKK
ABCAAB137441-100
Anti-PIP5K1A Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: PIP5K1A (phosphatidylinositol-4-phosphate 5-kinase, type I, alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR