Anti-GABRA1 Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB137436-50EA 6480 DKK
ABCAAB137436-50
Anti-GABRA1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: GABRA1 (gamma-aminobutyric acid (GABA) A receptor, alpha 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR