Anti-USP14 Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB137433-50EA 6580 DKK
ABCAAB137433-50
Anti-USP14 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: USP14 (ubiquitin specific peptidase 14 (tRNA-guanine transglycosylase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR