Anti-CYP2D6 Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB137426-50EA 4100.24 DKK
ABCAAB137426-50
Anti-CYP2D6 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: CYP2D6 (cytochrome P450, family 2, subfamily D, polypeptide 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR