Anti-WEE1 Rabbit Polyclonal Antibody
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB137377-100EA 4100.24 DKK
ABCAAB137377-100
Anti-WEE1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: WEE1 (WEE1 G2 checkpoint kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human