Anti-BCL7A Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB137362-100EA 3802.04 DKK
ABCAAB137362-100
Anti-BCL7A Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: BCL7A (B-cell CLL/lymphoma 7A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR