Anti-NRL Rabbit Polyclonal Antibody
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB137193-100EA 3802.04 DKK
ABCAAB137193-100
Anti-NRL Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: NRL (neural retina leucine zipper)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human