Anti-ALDH16A1 Rabbit Monoclonal Antibody
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB137082-100EA 6630 DKK
ABCAAB137082-100
Anti-ALDH16A1 Rabbit Monoclonal Antibody
Antistoffer
Type: Primary
Antigen: ALDH16A1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestil nu