Anti-MAPK1/3 Rabbit Monoclonal Antibody [clone: G15-B]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB136926-50EA 3951.14 DKK
ABCAAB136926-50
Anti-MAPK1/3 Rabbit Monoclonal Antibody [clone: G15-B]
Antistoffer
Type: Primary
Antigen: MAPK1/3 (mitogen-activated protein kinase 1/3)
Clonality: monoclonal
Clone: G15-B
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR