Anti-PKC beta 1 Rabbit Monoclonal Antibody [clone: A10-F]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB136917-50EA 6430 DKK
ABCAAB136917-50
Anti-PKC beta 1 Rabbit Monoclonal Antibody [clone: A10-F]
Antistoffer
Type: Primary
Antigen: PKC beta 1
Clonality: monoclonal
Clone: A10-F
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR