Anti-Gelsolin plasma Goat Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB136832-200EA 3802.04 DKK
ABCAAB136832-200
Anti-Gelsolin plasma Goat Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: Gelsolin plasma
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR