Anti-IgG Rabbit Polyclonal Antibody (Biotin)

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB136769-500EA 2236.49 DKK
ABCAAB136769-500
Anti-IgG Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Type: Secondary
Antigen: IgG (Dog) (dog IgG)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR