Anti-IgG Rabbit Polyclonal Antibody (Glucose Oxidase)
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB136719-500EA 3460 DKK
ABCAAB136719-500
Anti-IgG Rabbit Polyclonal Antibody (Glucose Oxidase)
Antistoffer
Type: Secondary
Antigen: IgG (Goat) (goat IgG)
Clonality: polyclonal
Clone:
Conjugation: Glucose Oxidase
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestil nu