Anti-MitoSciences None Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB136649-100EA 3988.41 DKK
ABCAAB136649-100
Anti-MitoSciences None Antibody
Antistoffer
Type: Primary
Antigen: MitoSciences
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: None
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR