Anti-HuR / ELAVL1 Mouse Monoclonal Antibody [clone: 4C8]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB136542-100EA 4098.45 DKK
ABCAAB136542-100
Anti-HuR / ELAVL1 Mouse Monoclonal Antibody [clone: 4C8]
Antistoffer
Type: Primary
Antigen: HuR / ELAVL1
Clonality: monoclonal
Clone: 4C8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR