Anti-Sulfotyrosine Mouse Monoclonal Antibody [clone: Sulfo-1C-A2]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB136481-30EA 6500 DKK
ABCAAB136481-30
Anti-Sulfotyrosine Mouse Monoclonal Antibody [clone: Sulfo-1C-A2]
Antistoffer
Type: Primary
Antigen: Sulfotyrosine
Clonality: monoclonal
Clone: Sulfo-1C-A2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR