Anti-B4GALNT1 Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB135902-100EA 3802.04 DKK
ABCAAB135902-100
Anti-B4GALNT1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: B4GALNT1 (beta-1,4-N-acetyl-galactosaminyl transferase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR