Anti-AGPAT3 Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB135887-100EA 6150 DKK
ABCAAB135887-100
Anti-AGPAT3 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: AGPAT3 (1-acylglycerol-3-phosphate O-acyltransferase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR