Anti-Influenza A virus (H5N1/HA1) None Antibody [clone: 2B7]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB135382-100EA 6450 DKK
ABCAAB135382-100
Anti-Influenza A virus (H5N1/HA1) None Antibody [clone: 2B7]
Antistoffer
Type: Primary
Antigen: Influenza A virus (H5N1/HA1)
Clonality:
Clone: 2B7
Conjugation: unconjugated
Epitope:
Host: None
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR