Anti-DCX Mouse Monoclonal Antibody [clone: 2G5]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB135349-100EA 6710 DKK
ABCAAB135349-100
Anti-DCX Mouse Monoclonal Antibody [clone: 2G5]
Antistoffer
Type: Primary
Antigen: DCX (doublecortin)
Clonality: monoclonal
Clone: 2G5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR