Anti-Nicotinic Acetylcholine Receptor alpha 1 Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB135272-100EA 6480 DKK
ABCAAB135272-100
Anti-Nicotinic Acetylcholine Receptor alpha 1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: Nicotinic Acetylcholine Receptor alpha 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR