Anti-CA9 Mouse Monoclonal Antibody [clone: MM0610-3B15]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB135159-100EA 6180 DKK
ABCAAB135159-100
Anti-CA9 Mouse Monoclonal Antibody [clone: MM0610-3B15]
Antistoffer
Type: Primary
Antigen: CA9 (carbonic anhydrase IX)
Clonality: monoclonal
Clone: MM0610-3B15
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR