Anti-PHOX2A Rabbit Monoclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB134939-100EA 3692.6 DKK
ABCAAB134939-100
Anti-PHOX2A Rabbit Monoclonal Antibody
Antistoffer
Type: Primary
Antigen: PHOX2A
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR