Anti-XAGE1A Goat Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB134805-50EA 3802.04 DKK
ABCAAB134805-50
Anti-XAGE1A Goat Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: XAGE1A (X antigen family, member 1A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR