Anti-LARP7 Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB134746-100EA 4100.24 DKK
ABCAAB134746-100
Anti-LARP7 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: LARP7 (La ribonucleoprotein domain family, member 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR