Anti-CCK Rabbit polyclonal antibody unconjugated

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB134713-100EA 6480 DKK
ABCAAB134713-100
Anti-CCK Rabbit polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: CCK (cholecystokinin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR