Anti-P2RX4 Goat Polyclonal Antibody
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB134559-200EA 4098.45 DKK
ABCAAB134559-200
Anti-P2RX4 Goat Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: P2RX4 (purinergic receptor P2X, ligand-gated ion channel, 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human