Anti-CD200 / OX2 Mouse Monoclonal Antibody (APC (Allophycocyanin) [clone: OX-104]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB134489-50EA 2236.49 DKK
ABCAAB134489-50
Anti-CD200 / OX2 Mouse Monoclonal Antibody (APC (Allophycocyanin) [clone: OX-104]
Antistoffer
Type: Primary
Antigen: CD200 / OX2
Clonality: monoclonal
Clone: OX-104
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR