Anti-ITGA2B Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MEM-06]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB134372-50EA 4100.24 DKK
ABCAAB134372-50
Anti-ITGA2B Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MEM-06]
Antistoffer
Type: Primary
Antigen: ITGA2B (integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigen CD41))
Clonality: monoclonal
Clone: MEM-06
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR