Anti-THY1 Mouse Monoclonal Antibody (Biotin) [clone: 5E10]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB134361-50EA 4100.24 DKK
ABCAAB134361-50
Anti-THY1 Mouse Monoclonal Antibody (Biotin) [clone: 5E10]
Antistoffer
Type: Primary
Antigen: THY1 (Thy-1 cell surface antigen)
Clonality: monoclonal
Clone: 50000000000
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR